Order Entry
China
Orders LinkContactUsLinkComponent
Section lifters

Section lifters

150 mm, 50 mm, 15 mm
Catalog # RSGA063.015
Supplier:   RSG
Section lifters
Catalog # RSGA063.015
Supplier:  RSG
Section lifters
Section lifters

150 mm, 50 mm, 15 mm

Price
Inquire For Price
Log in for availability
1 * 5 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Blade made of stainless steel, wooden handle.

  • Blade with excellent heat resistance
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    150 mm
  • Blade width:
    15 mm
  • Blade length:
    50 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.