Order Entry
China
Orders LinkContactUsLinkComponent
Spoon spatulas, flat

Spoon spatulas, flat

Double spoon spatula, flat endings, 180 mm, 10 mm
Catalog # RSGA038.180
Supplier:   RSG
Spoon spatulas, flat
Catalog # RSGA038.180
Supplier:  RSG
Spoon spatulas, flat
Spoon spatulas, flat

Double spoon spatula, flat endings, 180 mm, 10 mm

Price
Inquire For Price
Log in for availability
1 * 5 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Flat spoon spatulas made of stainless steel. Available singe or double ended.

Specifications
  • Description:
    Double spoon spatula, flat endings
  • Length:
    180 mm
  • Blade width:
    10 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.