Tissue capsules
White, 28×5 mm
SIMPM470
These capsules made of white PP can hold tissue during processing.
- Frosted area for writing sample ID
- Lid snaps securely on to the base
- Type:28×5 mm
- Colour:White
These capsules made of white PP can hold tissue during processing.