Order Entry
China
ContactUsLinkComponent
Clamp, 3-prong dual adjust, 18/10 steel, d=0-69 mm, d=13mm, l=127 mm, span width d independently adjustable by 2 regulating screws, offers a wide range of motion. prongs supplied with non-slip vinyl sleeves. 1 * 1 items
Catalog # BOCH5701
Supplier:  BOCHEM
Clamp, 3-prong dual adjust, 18/10 steel, d=0-69 mm, d=13mm, l=127 mm, span width d independently adjustable by 2 regulating screws, offers a wide range of motion. prongs supplied with non-slip vinyl sleeves. 1 * 1 items
Catalog # BOCH5701
Supplier:  BOCHEM