Order Entry
China
Orders LinkContactUsLinkComponent
 

VWR® AVS TITRINORM Buffer Solutions, 20 °C

10,00/7,00/4,00, ±0,02, Blue/green/red, Plastic bottle, 3×100 ml, 1
Catalog # VWRC83610.600
Supplier:   VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Catalog # VWRC83610.600
Supplier:  VWR Chemicals
CAS Number:  
VWR® AVS TITRINORM Buffer Solutions, 20 °C
VWR® AVS TITRINORM Buffer Solutions, 20 °C

10,00/7,00/4,00, ±0,02, Blue/green/red, Plastic bottle, 3×100 ml, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.

  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website

Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.

 : Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.

 : If significantly large volumes are required please contact your local Avantor sales office for a customised offer.

 : Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1
  • Colour:
    Blue/green/red
  • pH accuracy:
    ±0,02
  • Pk Info:
    3×100 ml
  • pH value:
    10,00/7,00/4,00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.