Sodium hydroxide 50% in aqueous solution, TECHNICAL
Plastic bottle, 5 L
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Catalog # VWRC28265.366
Supplier: VWR Chemicals
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1.53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Certificates
Search by Lot Number2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC28265.322_SPEC_EN.pdf P20_VWRC28265.366_SPEC_EN.pdf P20_VWRC28265.446_SPEC_EN.pdf spec_VWRC28265.322 spec_VWRC28265.366
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:47 - 51 %
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Cat. No.:VWRC28265.366
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



