2-Propanol ≥98%, TECHNICAL
Metal drum, 200 L
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Catalog # VWRC20922.557
Supplier: VWR Chemicals
CAS Number: 67-63-0
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
More
P20_VWRC20922.320_SPEC_EN.pdf P20_VWRC20922.364_SPEC_EN.pdf P20_VWRC20922.368_SPEC_EN.pdf P20_VWRC20922.411_SPEC_EN.pdf P20_VWRC20922.466_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pack type:Metal drum
- Pk:200 L
- Cat. No.:VWRC20922.557
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



