Order Entry
China
Orders LinkContactUsLinkComponent
Bossheads for frames, 90°

Bossheads for frames, 90°

13 mm, Tempered cast iron, chromed
  BOCH5363
 :   BOCHEM
Bossheads for frames, 90°
  BOCH5363
 :  BOCHEM
Bossheads for frames, 90°
Bossheads for frames, 90°

13 mm, Tempered cast iron, chromed

 
 
   
1 * 1 件
 

For rods up to 13 mm.

  • Angle: 90°
  • Heart screws
 
  • Spanning width:
    13 mm
  • Material:
    Tempered cast iron, chromed