Order Entry
China
ContactUsLinkComponent
Lid for wind-shielded rotors 75006606. For use with double blood pack bucket 75007617 (essential when using an old 6606 rotor with buckets 7617) 1 * 1 items
Catalog # HERA75007614
Supplier:  THERMO HERAUS
Lid for wind-shielded rotors 75006606. For use with double blood pack bucket 75007617 (essential when using an old 6606 rotor with buckets 7617) 1 * 1 items
Catalog # HERA75007614
Supplier:  THERMO HERAUS