Order Entry
China
Orders LinkContactUsLinkComponent
EDTA solutions
EDTA solutions
EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration, Self-contained dropping bottle, 1 L
Catalog # HACH20553
Supplier:   HACH
EDTA solutions
Catalog # HACH20553
Supplier:  HACH
CAS Number:  
EDTA solutions
EDTA solutions
EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration, Self-contained dropping bottle, 1 L
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 升
Product Details & Documents
Documents
Specifications
Specifications
  • Pack type:
    Self-contained dropping bottle
  • Pk:
    1 L
  • Description:
    EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration
Specifications