VWR® AVS TITRINORM Buffer Solutions, 20 °C
Product Details & Documents
Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.
- Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.
Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.
Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.
Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).
Documents
Certificates
按批号搜索100 ml plastic bottle for liquids DIN 28 (ZPACK13948098) 10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 250 ml Glass bottle for liquids DIN 28 (ZPACK13949250) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Packaging Information
P20_VWRC32031.297_SPEC_EN.pdf P20_VWRC32032.260_SPEC_EN.pdf P20_VWRC32032.291_SPEC_EN.pdf P20_VWRC32032.291_SPEC_PL.pdf P20_VWRC32033.294_SPEC_EN.pdf
Product specification
SDS_EN_CH_VWRC32031 SDS_EN_CH_VWRC32044 SDS_EN_CH_VWRC32045 SDS_EN_CH_VWRC32045.pdf SDS_EN_CH_VWRC32046
SDS
VWR® for Calibrations VWR® pH Buffer Standard Solutions.pdf
Product Literature
规格
规格
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:500 mL
- Colour:Colourless
- pH accuracy:±0,02
- pH value:9,22
- Accreditation:ISO 17034
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



