Order Entry
China
Orders LinkContactUsLinkComponent
Buffer powder pillows

Buffer powder pillows

10,01, ±0,02, Carbonate, Blue, 25 °C, 50
目录编号 HACH2227166
供应商:   HACH
Buffer powder pillows
目录编号 HACH2227166
供应商:  HACH
CAS号:  
Buffer powder pillows
Buffer powder pillows

10,01, ±0,02, Carbonate, Blue, 25 °C, 50

Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 50 件
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

 : The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

规格

规格

  • pH buffer type:
    Carbonate
  • Pk:
    50
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,02
  • pH value:
    10,01

规格