Order Entry
China
Orders LinkContactUsLinkComponent
Micro double spatulas

Micro double spatulas

130 mm, 40 mm, 8 mm
  BOCH3047
 :   BOCHEM
Micro double spatulas
  BOCH3047
 :  BOCHEM
Micro double spatulas
Micro double spatulas

130 mm, 40 mm, 8 mm

 
 
   
1 * 1 件
 

Nickel. Multipurpose spatulas for a wide range of uses.

  • With round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
 
  • Length:
    130 mm
  • Seq:
    02
  • Blade width:
    8 mm
  • Blade length:
    40 mm