Order Entry
China
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

185 mm, 50 mm, 9 mm
Catalog # BOCH3305
Supplier:   BOCHEM
Double spatulas, micro
Catalog # BOCH3305
Supplier:  BOCHEM
Double spatulas, micro
Double spatulas, micro

185 mm, 50 mm, 9 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents

Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Specifications
  • Length:
    185 mm
  • Blade width:
    9 mm
  • Blade length:
    50 mm