0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
General purpose hydrometer w/o thermometer, 0,600 to 0,800 g/cm subdiv. 0,002 g/cm 1 * 1 items
Catalog # BRND969510
Supplier:
Brand
Expand image
General purpose hydrometer w/o thermometer, 0,600 to 0,800 g/cm subdiv. 0,002 g/cm 1 * 1 items
Catalog # BRND969510
Supplier:
Brand