Order Entry
China
Orders LinkContactUsLinkComponent
 

Sampling siphons

Sampling siphon, PE, 175 ml, 750 mm
  BURK5387-0075
 :   BURKLE
Sampling siphons
  BURK5387-0075
 :  BURKLE
Sampling siphons
Sampling siphons

Sampling siphon, PE, 175 ml, 750 mm

 
 
   
1 * 1 件
 
   
 

Disposable plunging samplers acting like siphons.

  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler

If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.

 
  • Capacity:
    175 ml
  • Description:
    Sampling siphon
  • Length:
    750 mm
  • Material:
    PE
  • Seq:
    02

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.