Volumetric flasks, class B
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
- NS:24/29
- Capacity:1000 ml
- Seq:07
- Tolerance:0,80 ± ml