Order Entry
China
Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
50 mm
  USBE5516
Hoffmann pinch cocks
  USBE5516
Hoffmann pinch cocks
Hoffmann pinch cocks
50 mm
1 * 1 件
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
 : The values in the band width column refer to the clearance between the pinch cock legs.
  • Bandwidth:
    50 mm