Order Entry
China
Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
Hoffmann pinch cocks
25 mm
Catalog # USBE5513
Supplier:   USBECK LABORGERATE
Hoffmann pinch cocks
Catalog # USBE5513
Supplier:  USBECK LABORGERATE
Hoffmann pinch cocks
Hoffmann pinch cocks
25 mm
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
Ordering information: The values in the band width column refer to the clearance between the pinch cock legs.
Specifications
  • Bandwidth:
    25 mm