Order Entry
China
Orders LinkContactUsLinkComponent
Spoon spatulas, broad shape
Spoon spatulas, broad shape
120 mm, 23 mm
Catalog # USBE3331
Supplier:   USBECK LABORGERATE
Spoon spatulas, broad shape
Catalog # USBE3331
Supplier:  USBECK LABORGERATE
Spoon spatulas, broad shape
Spoon spatulas, broad shape
120 mm, 23 mm
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rigid spoon spatulas made of stainless steel. With spoon end and one flat end.
Specifications
  • Length:
    120 mm
  • Spoon size:
    23 mm