Modules, Nunc-Immuno™ LockWell™
Product Details & Documents
PS. Nunc-Immuno™ LockWell™ modules facilitate assays from large scale screening to single-well tests. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
- Each well locks in the frame at the same height to prevent jamming in automated systems
- Strips are easy to take out or put in the frame, without use of any special tools
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules; MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum; MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit website or contact your local Avantor sales office for further information.
Specifications
- No. of wells:96
- Colour:Clear
- Description:PolySorp™, C-StarWell™ 8×12
- Seq:07
Accessories
Product Information
- Catalog #AvailabilityPrice
- LockWell™ frame