Plates, Nunc-Immuno™ TSP (transferable solid phases)
Nunc-Immuno™ TSP lids, sterile, for hybridoma screening
Catalog # NUNC445497
Supplier: Thermo Fisher Scientific
Plates, Nunc-Immuno™ TSP (transferable solid phases)
Catalog # NUNC445497
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS. For simultaneous testing of analytes using ELISA technique. Also available sterile for screening of hybridoma cultures for antibody production.
- Uniform adsorption properties
- Plates have 96 round bottom wells with total surface area of 95 mm²
- Sterile plates supplied mounted in the tray
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains.
Colour: Clear
Colour: Clear
Specifications
- Description:Nunc-Immuno™ TSP lids, sterile, for hybridoma screening