Sample tubes
Product Details & Documents
These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.
- Tube made of PP
- Cap made of PE
- Self-standing base with graduation
- Leakproof screw cap
- Available sterile and non-sterile
- Functional temperature range: –90 to +121 °C
Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-in graduations make them easy to read.
Packaging: Please note that the non-sterile caps and tubes are package separately in packages of 100. Sterile tubes are supplied in packs of 25.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Ext.Ø:30 mm
- Version:sterile
- Capacity:50 ml
- Height:115 mm
- Cap colour:Green
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



