Order Entry
China
Orders LinkContactUsLinkComponent
Adapters, distilling heads
14/23 NS, 24/29 - 19/26 NS
  LENZ05028032
Adapters, distilling heads
  LENZ05028032
Adapters, distilling heads
Adapters, distilling heads
14/23 NS, 24/29 - 19/26 NS
1 * 1 件
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Includes a socket for a distillation thermometer of 55 mm fitted length and two cones at an angle of 75°.
  • Cone size:
    24/29 - 19/26 NS
  • Socket size:
    14/23 NS