Order Entry
China
Orders LinkContactUsLinkComponent
Separating funnels, Gilson
2000 ml, 29/32 NS
  LENZ04030173
Separating funnels, Gilson
  LENZ04030173
Separating funnels, Gilson
Separating funnels, Gilson
2000 ml, 29/32 NS
1 * 1 件
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml