Order Entry
China
Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
1000 ml, 29/32 NS
Catalog # LENZ04030070
Supplier:   Lenz Laborglas
Separating funnels, Gilson
Catalog # LENZ04030070
Supplier:  Lenz Laborglas
Separating funnels, Gilson
Separating funnels, Gilson
1000 ml, 29/32 NS
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml