Mercury 99.99+%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7439-97-6
- Formula:Hg
- MDL Number:MFCD00011035
- Molecular Weight:200.59 g/mol
- Merck Index:13,05925
- Boiling Pt:356.5 °C (1013 hPa)
- Melting Pt:–38.9 °C
- Density:13.54 g/cm³
- Storage Temperature:Ambient
- EINECS:231-106-7
- UN:2809
- ADR:8,III
Documents
Specifications
Specification Test Results
- Appearance:Silver white Metallic liquid
- Assay:≥99.990 %
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
- Silver (Ag):≤1 ppm
- Zinc (Zn):≤0.5 ppm
- Insoluble matter:≤0.002 % (in HNO3)
Specifications
- Pack type:Plastic bottle
- Pk:100 g



