Sulphamethazine 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Appearance: White to light beige Crystalline powder
Synonyms: 4,6-Dimethylsulphadiazine, Sulphadimidine, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide
- CAS Number:57-68-1
- Formula:C₁₂H₁₄N₄O₂S
- MDL Number:MFCD00006066
- Molecular Weight:278.33 g/mol
- Melting Pt:197…200 °C
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:197 to 200 °C
- Titration with HClO4:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



