Pipette tips, microvolume, Corning®
Round tip, 0,5 - 10 µl, -, Transparent, Eppendorf, other Ultra-Micropipettors, Bulk; 10000 tips/bag
Supplier: CORNING SCIENCE PRODUCTS
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Certificates
Search by Lot NumberSpecifications
- Packed:Bulk; 10000 tips/bag
- Sterile:-
- Volume:0,5 - 10 µl
- Seq:06
- Tip style:Round tip
- Tip colour:Transparent
- For use with:Eppendorf, other Ultra-Micropipettors