0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Insert 12mm Transwell-COL polystyrene with 3,0µm pore PTFE membrane collagen coated sterile 1 * 24 items
Catalog # CORN3494
Supplier:
CORNING SCIENCE PRODUCTS
Expand image
Insert 12mm Transwell-COL polystyrene with 3,0µm pore PTFE membrane collagen coated sterile 1 * 24 items
Catalog # CORN3494
Supplier:
CORNING SCIENCE PRODUCTS