山梨酸 99%
3401, Plastic bottle, 100 g
: 2,4-己二烯酸
- :110-44-1
- :CH₃CH=CHCH=CHCOOH
- :MFCD00002703
- :112.13 g/mol
- :13,08793
- :228 °C (1013 hPa)
- :132…135 °C
- :1,204 g/cm³ (20 °C)
- :127 °C (closed cup)
- :Ambient
- :203-768-7
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:132 to 135 °C
- Water:≤0.5 % (K.F.)
- Sulfated ash:≤0.2 %
- Titration with NaOH:98.5 to 101.5 % (On anhydrous substance)
- Arsenic (As):≤3 ppm
- Heavy metals (as Pb):≤10 ppm
- 包装类型:Plastic bottle
- 包装规格:100 g
- 顺序:3401



