Sorbic acid 99%
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
- CAS Number:110-44-1
- Formula:CH₃CH=CHCH=CHCOOH
- MDL Number:MFCD00002703
- Molecular Weight:112.13 g/mol
- Merck Index:13,08793
- Boiling Pt:228 °C (1013 hPa)
- Melting Pt:132…135 °C
- Density:1,204 g/cm³ (20 °C)
- Flash Pt:127 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:203-768-7
Documents
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:132 to 135 °C
- Water:≤0.5 % (K.F.)
- Sulfated ash:≤0.2 %
- Titration with NaOH:98.5 to 101.5 % (On anhydrous substance)
- Arsenic (As):≤3 ppm
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



