Cyclohexanone 99.8%, extra pure
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
Cyclohexanone 99.8%, extra pure
Catalog # ACRO111190250
Supplier: Thermo Fisher Scientific
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155 °C (760 mmHg)
- Melting Pt:–47 °C
- Density:0.947 g/cm³
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4520 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥99.75 %
- Color scale:≤20 APHA
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.005 %
- Manganese (Mn):≤0.002 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.05 %
- Insoluble matter:≤0.005 %
- Nitrogen compounds (as N):≤0.002 %
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Cat. No.:ACRO111190250
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health



