Pepsin
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Contains either Lactose or Maltodextrin excipient
- Assay: >=0.5 Ph. Eur. U/mg (Activity)
- Assay: Hemoglobin, pH 1.6, 25°C
- Infrared spectrum: Conforms
- Loss on drying: =<5.0 % (60°C, 4 h, 670 Pa) (P2O5)
- Microbial contamination: Passes test
- Sulfated ash: =<5.0 % (800°C)
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Cat. No.:ACRO417075000
- Form:White to light yellow powder
- Storage conditions:Refrigerator
- pH:3.0 to 5.0 (2 % in water)
- MDL No.:MFCD00081840
- Enzyme name:Pepsin
- CAS No.:9001-75-6
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



