Order Entry
China
Orders LinkContactUsLinkComponent
Slide boxes, PS
Slide boxes, PS
Grey, PS, 100, 230×187×35 mm
Catalog # BRND476000
Supplier:   BRAND
Slide boxes, PS
Catalog # BRND476000
Supplier:  BRAND
Slide boxes, PS
Slide boxes, PS
Grey, PS, 100, 230×187×35 mm
Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents
Grey polystyrene slide boxes.
Specifications
  • No. of slides:
    100
  • Colour:
    Grey
  • Material:
    PS
  • Seq:
    003
  • W×D×H:
    230×187×35 mm