Double spatulas
Double spatula ends, 180 mm, Green
BRND139815
: BRAND
PS, green. These impact resistant spatulas are available with double spatula ends or spatula and spoon combination.
- Colour:Green
- Description:Double spatula ends
- Length:180 mm