Order Entry
China
Orders LinkContactUsLinkComponent
Test tube racks, rectangular

Test tube racks, rectangular

24 (2×12), 20 mm, 310×60×90 mm
Catalog # BOCH8222
Supplier:   BOCHEM
Test tube racks, rectangular
Catalog # BOCH8222
Supplier:  BOCHEM
Test tube racks, rectangular
Test tube racks, rectangular

24 (2×12), 20 mm, 310×60×90 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents

Rack, made of stainless steel 18/10.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • For tube Ø:
    20 mm
  • No. of Holes:
    24 (2×12)
  • Seq:
    05
  • W×D×H:
    310×60×90 mm