Test tube racks, rectangular
12 (2×6), 20 mm, 164×60×90 mm
Catalog # BOCH8220
Supplier: BOCHEM
Jump to:
Product Details & Documents
Rack, made of stainless steel 18/10.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
Specifications
- For tube Ø:20 mm
- No. of Holes:12 (2×6)
- W×D×H:164×60×90 mm



