Test tube racks, rectangular
48 (4×12), 17 mm, 287×97×90 mm
Catalog # BOCH8214
Supplier: BOCHEM
Jump to:
Product Details & Documents
Rack, made of stainless steel 18/10.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
Specifications
- For tube Ø:17 mm
- No. of Holes:48 (4×12)
- W×D×H:287×97×90 mm



