Section lifters
150 mm, 50 mm, 15 mm
Catalog # BOCH3446
Supplier: BOCHEM
Jump to:
Product Details & Documents
Section lifters with flexible blade and handle made of stainless steel.
Specifications
- Length:150 mm
- Blade width:15 mm
- Blade length:50 mm



