(±)-1-Ethoxy-2-propanol 95%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
(±)-1-Ethoxy-2-propanol 95%
Catalog # ACRO259050010
Supplier: Thermo Fisher Scientific
CAS Number: 1569-02-4
Jump to:
Product Details & Documents
Synonyms: 1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether
- CAS Number:1569-02-4
- Formula:C₅H₁₂O₂
- MDL Number:MFCD00067050
- Molecular Weight:104.15 g/mol
- Boiling Pt:132 °C (760 mmHg)
- Melting Pt:–100 °C
- Density:0.897 g/cm³
- Flash Pt:42 °C
- Storage Temperature:Ambient
- EINECS:216-374-5
- UN:3271
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4050 to 1.4090 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥94 % Remark: balance 2-ethoxy-1-propanol
Specifications
- Pack type:Glass bottle
- Pk:1 L



