Lead(II) carbonate basic, extra pure
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Lead(II) carbonate basic, extra pure
Catalog # ACRO222710025
Supplier: Thermo Fisher Scientific
CAS Number: 1319-46-6
Jump to:
Product Details & Documents
Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
- CAS Number:1319-46-6
- Formula:(PbCO₃)₂·Pb(OH)₂
- MDL Number:MFCD00078155
- Molecular Weight:775.63 g/mol
- Melting Pt:400 °C (dec.)
- Density:6,06 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-290-6
- UN:2291
- ADR:6.1,III
- :Density: 6.06 g/cm³ (20 °C)
Documents
ACRO22271
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Titration Complexometric:≥75 % Pb
- Insoluble matter:≤0.05 % (in dilute acetic acid)
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



