Order Entry
China
Orders LinkContactUsLinkComponent
Buffer solutions in sachets

Buffer solutions in sachets

7,01, ±0,01, Sachet, 25×20 ml, 25
Catalog # HANNHI70007P
Supplier:   HANNA
Buffer solutions in sachets
Catalog # HANNHI70007P
Supplier:  HANNA
CAS Number:  
Buffer solutions in sachets

7,01, ±0,01, Sachet, 25×20 ml, 25

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 25 件
Product Details & Documents

For calibration of pH meters.

  • Colour coded label for easy pH buffer value identification
  • Ideal for field use due to portability
  • Light block packaging prevents oxidation from UV light

Traceable to NIST standard reference materials.

Documents

Specifications

Specifications

  • Pack type:
    Sachet
  • Pk:
    25
  • pH accuracy:
    ±0,01
  • Pk Info:
    25×20 ml
  • pH value:
    7,01

Specifications