Buffer solutions in sachets
4,01, ±0,01, Sachet, 25×20 ml, 25
Supplier: HANNA
Jump to:
Product Details & Documents
For calibration of pH meters.
- Colour coded label for easy pH buffer value identification
- Ideal for field use due to portability
- Light block packaging prevents oxidation from UV light
Traceable to NIST standard reference materials.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Sachet
- Pk:25
- pH accuracy:±0,01
- Pk Info:25×20 ml
- pH value:4,01



