Order Entry
China
ContactUsLinkComponent
VECTASPIN CENTRIFUGE TUBE FILTER, 400µL POLYSULPHONE 30K MOLECULAR WEIGHT CUTT OFF 100/PK 1 * 100 items
Catalog # WHAT6835-3001
Supplier:  CYTIVA WHATMAN
VECTASPIN CENTRIFUGE TUBE FILTER, 400µL POLYSULPHONE 30K MOLECULAR WEIGHT CUTT OFF 100/PK 1 * 100 items
Catalog # WHAT6835-3001
Supplier:  CYTIVA WHATMAN