Order Entry
China
ContactUsLinkComponent
VECTASPIN CENTRIFUGE TUBE FILTER, 400µL CELLULOSE ACETATE 20K MOLECULAR WEIGHT CUT OFF 100/PK 1 * 100 items
Catalog # WHAT6834-2001
Supplier:  CYTIVA WHATMAN
VECTASPIN CENTRIFUGE TUBE FILTER, 400µL CELLULOSE ACETATE 20K MOLECULAR WEIGHT CUT OFF 100/PK 1 * 100 items
Catalog # WHAT6834-2001
Supplier:  CYTIVA WHATMAN