Vanillin
Vanillin
Catalog # EHERC17900580
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1cc(C=O)ccc1O
- Inchi: InChI=1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3
- IUPAC: 4-hydroxy-3-methoxybenzaldehyde
Specifications
Specifications
- Description:Vanillin
- Cat. No.:EHERC17900580
- Formula:C₈H₈O₃
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Vanillin;Benzaldehyde;4-hydroxy-3-methoxy-;Vanillin (8CI);2-Methoxy-4-formylphenol;3-Methoxy-4-hydroxybenzaldehyde;4-Formyl-2-methoxyphenol;4-Hydroxy-3-methoxy-benzyldehyde;4-Hydroxy-3-methoxybenzaldehyde;4-Hydroxy-5-methoxybenzaldehyde;4-Hydroxy-m-anisaldehyde;H 0264;Lioxin;NPLC 0145;NSC 15351;NSC 403658;NSC 48383;Rhovanil;Vanillaldehyde;Vanillic aldehyde;Vanillum;m-Methoxy-p-hydroxybenzaldehyde;p-Hydroxy-m-methoxybenzaldehyde;p-Vanillin
- Molecular weight:152,15
- CAS No.:121-33-5
- Shipping conditions:RT



