D-Mannit
D-Mannit
Catalog # EHERC14752000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
- Inchi: InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1
- IUPAC: (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Specifications
Specifications
- Description:D-Mannit
- Cat. No.:EHERC14752000
- Formula:C₆H₁₄O₆
- Storage temperature:20 °C
- Synonym(s):Mannitol;d-Mannitol;D-Mannitol;Cordycepic acid (6CI,7CI);Mannitol;D- (8CI);Brightmoon;Bronchitol;Cerestar 16700;D-(-)-Mannitol;D-Mannit;Diosmol;E 421;Isotol;Manicol;Maniton S;Manna sugar;Mannidex;Mannidex 16700;Mannigen;Mannistol;Mannit;Mannit 60;Mannit P;Mannit S;Mannite;Mannitol 35;Mannitol 60;Mannitol P 60;Mannitol SD 200;Mannitolum;Mannogem 2080;Mannogem EZ;Marine Crystal;Nonpareil 108;Nonpareil 108(200);Orocell 200;Osmitrol;Osmosal;Parteck 300;Parteck Delta M;Parteck M 100;Parteck M 200;Parteck M 300;Partek M;Pearlitol;Pearlitol 100;Pearlitol 100SD;Pearlitol 160C;Pearlitol 2005D;Pearlitol 200SD;Pearlitol 25C;Pearlitol 50C;Pearlitol SD 100;Perlitol SD 200;Perteck M;PharmMannidex;PharmMannidex 16701;Resectisol;Lactitol Monohydrate Impurity C;Sorbitol Impurity A;myo-Inositol Impurity A;Lactitol Impurity C
- Molecular weight:182,17
- CAS No.:69-65-8
- Shipping conditions:RT
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



