D-Isoascorbic acid
D-Isoascorbic acid
Catalog # EHERC14379000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC[C@@H](O)[C@H]1OC(=O)C(=C1O)O
- Inchi: InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5-/m1/s1
- IUPAC: (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Specifications
Specifications
- Description:D-Isoascorbic acid
- Cat. No.:EHERC14379000
- Formula:C₆H₈O₆
- Storage temperature:20 °C
- Synonym(s):(5R)-5-[(1R)-1,2-Dihydroxyethyl]-3,4-dihydroxyfuran-2(5H)-one (D-Isoascorbic Acid);Ascorbic Acid Impurity F;Sodium Ascorbate Impurity F
- Molecular weight:176,12
- CAS No.:89-65-6
- Shipping conditions:RT



