Order Entry
China
Orders LinkContactUsLinkComponent
Acetamide

Acetamide

Acetamide
Catalog # EHERC10011800
Supplier:   EHRENSTORFER
Acetamide
Catalog # EHERC10011800
Supplier:  EHRENSTORFER
CAS Number:  
Acetamide
Acetamide

Acetamide

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 0,25 克
Product Details & Documents
  • SMILES: CC(=O)N
  • Inchi: InChI=1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)
  • IUPAC: acetamide
Specifications

Specifications

  • Description:
    Acetamide
  • Cat. No.:
    EHERC10011800
  • Formula:
    C₂H₅NO
  • Storage temperature:
    20 °C
  • Hazard class description:
    GHS08
  • GHS signal:
    Danger
  • Synonym(s):
    Acetamide;Acetic Acid Amide;Acetimidic Acid;Ethanamide;Ethanimidic Acid;Methanecarboxamide;NSC 25945;
  • Molecular weight:
    59,07
  • CAS No.:
    60-35-5
  • Shipping conditions:
    RT

Specifications